Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 18 - 19 | 2531 | 61 | |
GenBank mRNA | AB287295.1 | - | 18 - 19 | 2256 | 61 | |
AK141888.1 | - | 12 - 13 | 2978 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
