Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 26 - 27 | 4118 | 105 | |
GenBank mRNA | AB287295.1 | - | 26 - 27 | 3843 | 105 | |
AK088840.1 | - | 1 - 2 | 47 | 105 | ||
BC094345.1 | - | 7 - 8 | 1235 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
