Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 27 - 28 | 4239 | 104 | |
GenBank mRNA | AB287295.1 | - | 27 - 28 | 3964 | 104 | |
AK088840.1 | - | 2 - 3 | 168 | 104 | ||
BC094345.1 | - | 8 - 9 | 1356 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
