Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 31 - 32 | 4757 | 55 | |
GenBank mRNA | AB287295.1 | - | 31 - 32 | 4482 | 55 | |
AK088840.1 | - | 6 - 7 | 686 | 55 | ||
BC094345.1 | - | 12 - 13 | 1874 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
