Assay Details
Gene Symbol: Svop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026805.1 | NP_081081.1 | 1 - 2 | 230 | 65 | |
GenBank mRNA | AK047655.1 | - | 1 - 2 | 221 | 65 | |
AK047764.1 | - | 1 - 2 | 231 | 65 | ||
AK053833.1 | - | 1 - 2 | 221 | 65 | ||
AK082744.1 | - | 1 - 2 | 232 | 65 | ||
AK160770.1 | - | 1 - 2 | 221 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK047764.1, AK053833.1, AK082744.1, NM_026805.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
