Assay Details
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198109.4 | NP_932777.2 | 5 - 6 | 425 | 64 | |
GenBank mRNA | AB099287.1 | - | 5 - 6 | 460 | 64 | |
AK089308.1 | - | 5 - 6 | 429 | 64 | ||
AK155557.1 | - | 5 - 6 | 450 | 64 | ||
AK165380.1 | - | 4 - 5 | 390 | 64 | ||
AK170787.1 | - | 5 - 6 | 416 | 64 | ||
AK171210.1 | - | 5 - 6 | 474 | 64 | ||
AK171556.1 | - | 5 - 6 | 440 | 64 | ||
AK173144.1 | - | 5 - 6 | 447 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
