Assay Details
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010630.2 | NP_034760.2 | 10 - 11 | 1139 | 72 | |
GenBank mRNA | AK144304.1 | - | 11 - 12 | 1298 | 72 | |
D49545.1 | - | 10 - 11 | 1215 | 72 | ||
U92949.1 | - | 10 - 11 | 1129 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
