Assay Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 23 - 24 | 2795 | 75 | |
NM_053193.2 | NP_444423.1 | 23 - 24 | 2795 | 75 | ||
GenBank mRNA | AF322193.1 | - | 23 - 24 | 2766 | 75 | |
AK149914.1 | - | 16 - 17 | 1794 | 75 | ||
AK155806.1 | - | 23 - 24 | 2717 | 75 | ||
BC011458.1 | - | 19 - 20 | 2231 | 75 | ||
BC049941.1 | - | 23 - 24 | 2617 | 75 | ||
BC056388.1 | - | 23 - 24 | 2795 | 6854851 | 75 | |
BC057945.1 | - | 8 - 9 | 1126 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
