Assay Details
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030699.2 | NP_109624.1 | 6 - 7 | 2343 | 63 | |
GenBank mRNA | AB038667.1 | - | 6 - 7 | 2120 | 63 | |
AF475069.1 | - | 5 - 6 | 1248 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
