Assay Details
Gene Symbol: Eif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133916.2 | NP_598677.1 | 4 - 5 | 900 | 57 | |
GenBank mRNA | AK033722.1 | - | 4 - 5 | 900 | 57 | |
AK089978.1 | - | 4 - 5 | 888 | 57 | ||
AK167618.1 | - | 4 - 5 | 887 | 57 | ||
AK170938.1 | - | 4 - 5 | 900 | 57 | ||
BC007175.1 | - | 3 - 4 | 359 | 57 | ||
BC023767.1 | - | 4 - 5 | 871 | 5343631 | 57 | |
BC031704.1 | - | 4 - 5 | 877 | 4023277 | 57 | |
BC051065.1 | - | 4 - 5 | 860 | 5343631 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
