Assay Details
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081396.2 | NP_001074865.2 | 5 - 6 | 788 | 75 | |
NM_001167679.1 | NP_001161151.1 | 5 - 6 | 788 | 75 | ||
GenBank mRNA | AB185844.1 | - | 5 - 6 | 756 | 75 | |
AK084542.1 | - | 4 - 5 | 552 | 75 | ||
AK141805.1 | - | 5 - 6 | 658 | 75 | ||
AK154264.1 | - | 5 - 6 | 753 | 75 | ||
AK161403.1 | - | 4 - 5 | 605 | 75 | ||
AK171024.1 | - | 5 - 6 | 753 | 75 | ||
BC050039.1 | - | 5 - 6 | 734 | 30016266 | 75 | |
BC066830.1 | - | 5 - 6 | 720 | 30463017 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
