Assay Details
Gene Symbol: Srrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109909.1 | NP_001103379.1 | 14 - 15 | 2094 | 93 | |
NM_001109910.1 | NP_001103380.1 | 14 - 15 | 2094 | 93 | ||
NM_031405.2 | NP_113582.1 | 14 - 15 | 2094 | 93 | ||
GenBank mRNA | AK144300.1 | - | 14 - 15 | 2086 | 93 | |
AK150310.1 | - | 3 - 4 | 400 | 93 | ||
AK152710.1 | - | 3 - 4 | 400 | 93 | ||
BC019117.1 | - | 5 - 6 | 636 | 93 | ||
BC066831.1 | - | 14 - 15 | 2053 | 30471339 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK150310.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
