Assay Details
Gene Symbol: N4bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024917.1 | NP_001020088.1 | 10 - 11 | 2144 | 74 | |
GenBank mRNA | AK048616.1 | - | 5 - 6 | 1400 | 74 | |
AK173167.1 | - | 8 - 9 | 1855 | 74 | ||
BC046458.1 | - | 1 - 2 | 124 | 74 | ||
BC054549.1 | - | 10 - 11 | 2144 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
