Assay Details
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010298.6 | NP_034428.2 | 2 - 3 | 295 | 94 | |
GenBank mRNA | AK083251.1 | - | 1 - 2 | 149 | 94 | |
AM420316.1 | - | 1 - 2 | 128 | 94 | ||
BC037605.1 | - | 2 - 3 | 263 | 5361798 | 94 | |
U09399.1 | - | 2 - 3 | 204 | 94 | ||
X81202.1 | - | 1 - 2 | 87 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
