Assay Details
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081137.1 | NP_001074606.1 | 18 - 19 | 2215 | 119 | |
GenBank mRNA | EU937530.1 | - | 19 - 20 | 2278 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
