Assay Details
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081137.1 | NP_001074606.1 | 43 - 44 | 5076 | 77 | |
GenBank mRNA | EU937530.1 | - | 44 - 45 | 5139 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
