Assay Details
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081137.1 | NP_001074606.1 | 5 - 6 | 607 | 74 | |
GenBank mRNA | EU937530.1 | - | 6 - 7 | 670 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
