Assay Details
Gene Symbol: Smc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133786.3 | NP_598547.1 | 15 - 16 | 68 | ||
GenBank mRNA | AJ534940.2 | - | 14 - 15 | 68 | ||
AK048650.1 | - | 10 - 11 | 68 | |||
BC061481.1 | - | 15 - 16 | 6512832 | 68 | ||
BC062939.1 | - | 15 - 16 | 6841276 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
