Assay Details
Gene Symbol: Son | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178880.4 | NP_849211.3 | 7 - 8 | 6908 | 81 | |
GenBank mRNA | AK122418.1 | - | 5 - 6 | 1269 | 81 | |
BC145121.1 | - | 7 - 8 | 6850 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
