Assay Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013752.3 | NP_038780.3 | 14 - 15 | 2284 | 88 | |
GenBank mRNA | AB016988.1 | - | 14 - 15 | 2206 | 88 | |
AF076687.1 | - | 14 - 15 | 2278 | 88 | ||
AF092840.1 | - | 14 - 15 | 2267 | 88 | ||
AK132302.1 | - | 14 - 15 | 2326 | 88 | ||
AK159790.1 | - | 14 - 15 | 2290 | 88 | ||
AK160614.1 | - | 14 - 15 | 2251 | 88 | ||
BC003719.1 | - | 14 - 15 | 2293 | 88 | ||
BC044773.2 | - | 14 - 15 | 2283 | 5067851 | 88 | |
BC055061.1 | - | 14 - 15 | 2276 | 6528890 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
