Assay Details
Gene Symbol: Pls3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166453.1 | NP_001159925.1 | 11 - 12 | 1396 | 61 | |
NM_001166454.2 | NP_001159926.1 | 11 - 12 | 1391 | 61 | ||
NM_145629.2 | NP_663604.1 | 11 - 12 | 1510 | 61 | ||
GenBank mRNA | AB182243.1 | - | 10 - 11 | 1265 | 61 | |
AK146087.1 | - | 11 - 12 | 1321 | 61 | ||
AK146455.1 | - | 11 - 12 | 1313 | 61 | ||
AK160501.1 | - | 11 - 12 | 1339 | 61 | ||
BC005459.1 | - | 11 - 12 | 1471 | 3495462 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
