Assay Details
Gene Symbol: Pls3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166453.1 | NP_001159925.1 | 13 - 14 | 1639 | 73 | |
NM_001166454.2 | NP_001159926.1 | 13 - 14 | 1634 | 73 | ||
NM_145629.2 | NP_663604.1 | 13 - 14 | 1753 | 73 | ||
GenBank mRNA | AB182243.1 | - | 12 - 13 | 1508 | 73 | |
AK146087.1 | - | 13 - 14 | 1564 | 73 | ||
AK146455.1 | - | 13 - 14 | 1556 | 73 | ||
AK160501.1 | - | 13 - 14 | 1582 | 73 | ||
AK165540.1 | - | 13 - 14 | 1641 | 73 | ||
BC005459.1 | - | 13 - 14 | 1714 | 3495462 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
