Assay Details
Gene Symbol: Pls3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166453.1 | NP_001159925.1 | 5 - 6 | 633 | 81 | |
NM_001166454.2 | NP_001159926.1 | 5 - 6 | 628 | 81 | ||
NM_145629.2 | NP_663604.1 | 5 - 6 | 747 | 81 | ||
GenBank mRNA | AB182243.1 | - | 4 - 5 | 502 | 81 | |
AK146455.1 | - | 5 - 6 | 550 | 81 | ||
AK160501.1 | - | 5 - 6 | 576 | 81 | ||
BC005459.1 | - | 5 - 6 | 708 | 3495462 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
