Assay Details
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081149.1 | NP_001074618.1 | 17 - 18 | 3855 | 104 | |
GenBank mRNA | AK082463.1 | - | 17 - 18 | 3858 | 104 | |
AK132474.1 | - | 16 - 17 | 3763 | 104 | ||
AK143408.1 | - | 17 - 18 | 3846 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
