Assay Details
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 10 - 11 | 908 | 80 | |
GenBank mRNA | AF263345.1 | - | 10 - 11 | 915 | 80 | |
AK088364.1 | - | 10 - 11 | 930 | 80 | ||
AK133419.1 | - | 10 - 11 | 909 | 80 | ||
AK184114.1 | - | 1 - 2 | 77 | 80 | ||
AK211462.1 | - | 2 - 3 | 158 | 80 | ||
BC003477.1 | - | 10 - 11 | 898 | 80 | ||
BC008102.1 | - | 10 - 11 | 918 | 3966319 | 80 | |
D42068.1 | - | 10 - 11 | 900 | 80 | ||
U19520.1 | - | 10 - 11 | 922 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
