Assay Details
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 6 - 7 | 435 | 84 | |
GenBank mRNA | AF263345.1 | - | 6 - 7 | 442 | 84 | |
AK088364.1 | - | 6 - 7 | 457 | 84 | ||
AK133419.1 | - | 6 - 7 | 436 | 84 | ||
AK182048.1 | - | 2 - 3 | 162 | 84 | ||
BC003477.1 | - | 6 - 7 | 425 | 84 | ||
BC008102.1 | - | 6 - 7 | 445 | 3966319 | 84 | |
D42068.1 | - | 6 - 7 | 427 | 84 | ||
U19520.1 | - | 6 - 7 | 449 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
