Assay Details
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146131.2 | NP_666243.1 | 8 - 9 | 847 | 63 | |
GenBank mRNA | AK028754.1 | - | 8 - 9 | 808 | 63 | |
AK028858.1 | - | 8 - 9 | 845 | 63 | ||
AK159422.1 | - | 8 - 9 | 834 | 63 | ||
AK159448.1 | - | 8 - 9 | 834 | 63 | ||
AK159633.1 | - | 8 - 9 | 824 | 63 | ||
AK160103.1 | - | 8 - 9 | 828 | 63 | ||
AK170318.1 | - | 8 - 9 | 803 | 63 | ||
BC026838.1 | - | 7 - 8 | 782 | 4460993 | 63 | |
BC027771.1 | - | 8 - 9 | 817 | 5344900 | 63 | |
BC034200.1 | - | 8 - 9 | 788 | 4986595 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
