Assay Details
Gene Symbol: 4930478P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046060.1 | 1 - 2 | 1087 | 110 | ||
GenBank mRNA | AK016658.1 | - | 1 - 2 | 1088 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
