Assay Details
Gene Symbol: Rgs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163512.1 | NP_001156984.1 | 2 - 3 | 318 | 90 | |
NM_173402.2 | NP_775578.2 | 5 - 6 | 2365 | 90 | ||
GenBank mRNA | AK004813.2 | - | 3 - 4 | 271 | 90 | |
AK014569.1 | - | 2 - 3 | 320 | 90 | ||
AK145890.1 | - | 5 - 6 | 2313 | 90 | ||
AK161129.1 | - | 2 - 3 | 319 | 90 | ||
BC040396.1 | - | 5 - 6 | 2362 | 4461061 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
