Assay Details
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030721.2 | NP_109646.2 | 11 - 12 | 1366 | 122 | |
GenBank mRNA | AK028818.1 | - | 11 - 12 | 1787 | 122 | |
AK053960.1 | - | 11 - 12 | 1363 | 122 | ||
AK128922.1 | - | 11 - 12 | 1326 | 122 | ||
AK154112.1 | - | 10 - 11 | 1457 | 122 | ||
AK171598.1 | - | 1349 | 122 | |||
BC044725.2 | - | 11 - 12 | 1386 | 5374281 | 122 | |
BC055019.1 | - | 11 - 12 | 1366 | 4162223 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
