Assay Details
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030721.2 | NP_109646.2 | 17 - 18 | 2082 | 134 | |
GenBank mRNA | AJ278430.1 | - | 17 - 18 | 2115 | 134 | |
AK028818.1 | - | 17 - 18 | 2503 | 134 | ||
AK053960.1 | - | 17 - 18 | 2079 | 134 | ||
AK128922.1 | - | 17 - 18 | 2042 | 134 | ||
AK154112.1 | - | 16 - 17 | 2173 | 134 | ||
AK171598.1 | - | 2065 | 134 | |||
AK211732.1 | - | 2 - 3 | 129 | 134 | ||
BC044725.2 | - | 17 - 18 | 2102 | 5374281 | 134 | |
BC055019.1 | - | 17 - 18 | 2082 | 4162223 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
