Assay Details
Gene Symbol: Plrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016784.3 | NP_058064.2 | 12 - 13 | 1254 | 120 | |
GenBank mRNA | AF044334.1 | - | 11 - 12 | 1146 | 120 | |
AK075993.1 | - | 12 - 13 | 1242 | 120 | ||
AK076007.1 | - | 12 - 13 | 1232 | 120 | ||
AK077197.1 | - | 12 - 13 | 1255 | 120 | ||
BC006750.1 | - | 12 - 13 | 1224 | 3709134 | 120 | |
BC052895.1 | - | 12 - 13 | 1212 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
