Assay Details
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021369.2 | NP_067344.2 | 2 - 3 | 457 | 72 | |
GenBank mRNA | AK149262.1 | - | 2 - 3 | 398 | 72 | |
AY574265.1 | - | 2 - 3 | 215 | 72 | ||
BC031985.1 | - | 2 - 3 | 457 | 4501558 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
