Assay Details
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 4 - 5 | 482 | 131 | |
GenBank mRNA | AJ344447.1 | - | 4 - 5 | 482 | 131 | |
AK171707.1 | - | 4 - 5 | 406 | 131 | ||
BC066173.1 | - | 4 - 5 | 435 | 30049516 | 131 | |
DV651307.1 | - | 4 - 5 | 512 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
