Assay Details
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 5 - 6 | 629 | 62 | |
GenBank mRNA | AJ344447.1 | - | 5 - 6 | 629 | 62 | |
AK016254.1 | - | 3 - 4 | 309 | 62 | ||
AK171707.1 | - | 5 - 6 | 553 | 62 | ||
BC066173.1 | - | 5 - 6 | 582 | 30049516 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
