Assay Details
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 6 - 7 | 743 | 106 | |
GenBank mRNA | AJ344447.1 | - | 6 - 7 | 743 | 106 | |
AK016254.1 | - | 4 - 5 | 423 | 106 | ||
AK171707.1 | - | 6 - 7 | 667 | 106 | ||
BC066173.1 | - | 6 - 7 | 696 | 30049516 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
