Assay Details
Gene Symbol: Gnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153547.6 | NP_705775.2 | 7 - 8 | 825 | 91 | |
NR_104298.1 | 7 - 8 | 777 | 91 | |||
GenBank mRNA | AK077523.1 | - | 7 - 8 | 756 | 91 | |
AY181025.1 | - | 7 - 8 | 683 | 91 | ||
AY185498.1 | - | 7 - 8 | 683 | 91 | ||
BC018560.1 | - | 7 - 8 | 664 | 91 | ||
BC037996.1 | - | 7 - 8 | 685 | 5052858 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
