Assay Details
Gene Symbol: 4932441J04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015588.2 | 4 - 5 | 396 | 87 | ||
GenBank mRNA | AK039574.1 | - | 4 - 5 | 397 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 10440 | Gm10440 |
D130065D13, ENSMUSG00000072936 |
-3.2mb | Anti-sense | Chr. 5 - 54349992 - 54358542 on NCBI Build GRCm38 |
Downstream | protocadherin 7 | Pcdh7 | 115.0kb | Anti-sense | Chr. 5 - 57716456 - 58133235 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2285817 | AK044558.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2285812 | AK039574.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2285812 | NR_015588.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809586 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49228 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162671 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826015 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896634 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106133 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907357 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730595 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798608 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715477 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AB041034.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AB042745.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AF261944.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AK050371.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AK149398.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AK149455.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | AK149492.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | BC021378.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413018 | NM_015760.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116500 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103450 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171797 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2285817 | AK044558.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05581826 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05581865 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05581840 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05581857 | - | - |
Whole Mouse Genome G4122A | A_52_P660363 | AK039574.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P660363 | NR_015588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P94358 | AK034221.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P94358 | AK044558.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P94358 | AK030094.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128528 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05581865 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05581840 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720085 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04481892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719828 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719848 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720258 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04482130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04611298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719872 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720135 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720036 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04860083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720014 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04153663 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719861 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04261846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719854 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719842 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04608107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720120 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719858 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04749008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720197 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720119 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720176 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719834 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04230526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04860092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719856 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720021 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04772656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04261851 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720062 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04338017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04936442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720172 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04917428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719849 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04608074 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04860099 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719830 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04199473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23719835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720056 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04261844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04584614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23720270 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1436563_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436563_at | - | - |
Mouse Genome 430 2.0 Array | 1436563_PM_at | - | - |
Mouse Expression Set 430 | 1436563_PM_at | - | - |
Mouse Expression Set 430 | 1436563_at | - | - |
Murine Genome U74v2 Set | 170556_at | - | - |
Murine Genome U74v2 Set | 170556_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1436563_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1436563_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436563_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436563_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102970070 | - | - |
