Assay Details
Gene Symbol: Znf512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164597.1 | NP_001158069.1 | 6 - 7 | 1238 | 71 | |
GenBank mRNA | AK129308.1 | - | 6 - 7 | 1238 | 71 | |
BC056460.1 | - | 4 - 5 | 1225 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
