Assay Details
Gene Symbol: Pitpnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008851.4 | NP_032877.1 | 7 - 8 | 1733 | 104 | |
NR_075078.1 | 8 - 9 | 1427 | 104 | |||
GenBank mRNA | AF006467.1 | - | 8 - 9 | 1308 | 104 | |
AK155377.1 | - | 8 - 9 | 1383 | 104 | ||
BC044893.1 | - | 7 - 8 | 1238 | 4481550 | 104 | |
BC048150.1 | - | 7 - 8 | 1387 | 5706382 | 104 | |
Y08922.1 | - | 7 - 8 | 1734 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
