Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400002539.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002539.mm.1 |
NM_001199962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002539.mm.1 |
NM_001199962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002542.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002542.mm.1 |
NM_001199962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002542.mm.1 |
NM_001199962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001164046.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001164046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001199959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001199960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001199962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001199962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001199963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002549.mm.1 |
NM_001199963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002642.mm.1 |
TC0400000413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002642.mm.1 |
NM_001199960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002642.mm.1 |
NM_001199966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000047.mm.1 |
TC4_JH584293_random00000007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000047.mm.1 |
NM_001199961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000047.mm.1 |
NM_001267702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000045.mm.1 |
TC4_JH584293_random00000007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000045.mm.1 |
NM_001199961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000045.mm.1 |
NM_001267702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002643.mm.1 |
TC0400000413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002643.mm.1 |
NM_001199960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002643.mm.1 |
NM_001199961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002643.mm.1 |
NM_001199966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000006.mm.1 |
TC4_JH584294_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000006.mm.1 |
NM_001267703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000006.mm.1 |
NM_001267704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002538.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002538.mm.1 |
NM_001199962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002538.mm.1 |
NM_001199962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002545.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002545.mm.1 |
NM_001199960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002545.mm.1 |
NM_001199963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002545.mm.1 |
NM_001199963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020069.mm.1 |
TC0400002480.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020069.mm.1 |
NM_001048179.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020069.mm.1 |
NM_001048179.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020069.mm.1 |
NM_011336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020069.mm.1 |
NM_011336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000002.mm.1 |
TC4_JH584294_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000002.mm.1 |
NM_001267703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002546.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002546.mm.1 |
NM_001199960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002546.mm.1 |
NM_001199962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002546.mm.1 |
NM_001199962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002546.mm.1 |
NM_001199963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002546.mm.1 |
NM_001199963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000036.mm.1 |
TC4_JH584293_random00000007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000036.mm.1 |
NM_001199961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000036.mm.1 |
NM_001199965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000036.mm.1 |
NM_001267702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002646.mm.1 |
TC0400000413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002646.mm.1 |
NM_001199959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002646.mm.1 |
NM_001199960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002646.mm.1 |
NM_001199961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002646.mm.1 |
NM_001199966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000001.mm.1 |
TC4_JH584294_random00000001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584294_random00000001.mm.1 |
NM_001267703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002540.mm.1 |
TC0400000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002540.mm.1 |
NM_001199962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002540.mm.1 |
NM_001199962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000040.mm.1 |
TC4_JH584293_random00000007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000040.mm.1 |
NM_001199961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000040.mm.1 |
NM_001267702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000044.mm.1 |
TC4_JH584293_random00000007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000044.mm.1 |
NM_001199961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR4_JH584293_random00000044.mm.1 |
NM_001267702.1 |
Refseq |