Assay Details
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144517.4 | NP_653100.1 | 12 - 13 | 1028 | 112 | |
GenBank mRNA | AK076128.1 | - | 12 - 13 | 976 | 112 | |
AK083084.1 | - | 12 - 13 | 999 | 112 | ||
AK184122.1 | - | 5 - 6 | 324 | 112 | ||
AK217135.1 | - | 5 - 6 | 332 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
