Assay Details
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144517.4 | NP_653100.1 | 2 - 3 | 305 | 123 | |
GenBank mRNA | AK012836.1 | - | 2 - 3 | 257 | 123 | |
AK076128.1 | - | 2 - 3 | 253 | 123 | ||
BC020138.1 | - | 2 - 3 | 264 | 3983701 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
