Assay Details
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144517.4 | NP_653100.1 | 6 - 7 | 559 | 95 | |
GenBank mRNA | AK012836.1 | - | 6 - 7 | 511 | 95 | |
AK076128.1 | - | 6 - 7 | 507 | 95 | ||
AK083084.1 | - | 6 - 7 | 530 | 95 | ||
BC020138.1 | - | 6 - 7 | 518 | 3983701 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
