Assay Details
Gene Symbol: Sel1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172710.3 | NP_766298.2 | 6 - 7 | 1324 | 81 | |
GenBank mRNA | AK030487.1 | - | 6 - 7 | 1189 | 81 | |
AK037220.1 | - | 6 - 7 | 1188 | 81 | ||
AK122362.1 | - | 6 - 7 | 1324 | 81 | ||
BC094607.1 | - | 5 - 6 | 869 | 6837048 | 81 | |
BC117915.1 | - | 5 - 6 | 825 | 40097358 | 81 | |
BC117916.1 | - | 5 - 6 | 827 | 40097359 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
