Assay Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008069.4 | NP_032095.1 | 6 - 7 | 677 | 113 | |
GenBank mRNA | BC130258.1 | - | 6 - 7 | 681 | 40142758 | 113 |
BC157920.1 | - | 6 - 7 | 743 | 9007350 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
