Assay Details
Gene Symbol: Camsap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163749.1 | NP_001157221.1 | 9 - 10 | 1494 | 71 | |
NM_027171.3 | NP_081447.2 | 9 - 10 | 1491 | 71 | ||
GenBank mRNA | AK081728.1 | - | 9 - 10 | 1266 | 71 | |
AK220401.1 | - | 5 - 6 | 636 | 71 | ||
BC043033.1 | - | 1 - 2 | 109 | 71 | ||
BC043081.1 | - | 9 - 10 | 1150 | 71 | ||
BC048787.1 | - | 9 - 10 | 1139 | 6489361 | 71 | |
KC426930.1 | - | 11 - 12 | 1213 | 71 | ||
KC426931.1 | - | 11 - 12 | 1210 | 71 | ||
KC426932.1 | - | 10 - 11 | 1180 | 71 | ||
KC426933.1 | - | 10 - 11 | 1177 | 71 | ||
KC426934.1 | - | 10 - 11 | 1165 | 71 | ||
KC426935.1 | - | 10 - 11 | 1162 | 71 | ||
KC426936.1 | - | 9 - 10 | 1132 | 71 | ||
KC426937.1 | - | 9 - 10 | 1129 | 71 | ||
KC426938.1 | - | 11 - 12 | 1213 | 71 | ||
KC426939.1 | - | 11 - 12 | 1210 | 71 | ||
KC426940.1 | - | 10 - 11 | 1180 | 71 | ||
KC426941.1 | - | 10 - 11 | 1177 | 71 | ||
KC426942.1 | - | 10 - 11 | 1165 | 71 | ||
KC426943.1 | - | 10 - 11 | 1162 | 71 | ||
KC426944.1 | - | 9 - 10 | 1132 | 71 | ||
KC426945.1 | - | 9 - 10 | 1129 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
