Assay Details
Gene Symbol: Camsap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163749.1 | NP_001157221.1 | 10 - 11 | 1562 | 73 | |
NM_027171.3 | NP_081447.2 | 10 - 11 | 1559 | 73 | ||
GenBank mRNA | AK081728.1 | - | 10 - 11 | 1334 | 73 | |
AK220401.1 | - | 6 - 7 | 704 | 73 | ||
BC043033.1 | - | 2 - 3 | 177 | 73 | ||
BC043081.1 | - | 10 - 11 | 1218 | 73 | ||
BC048787.1 | - | 10 - 11 | 1207 | 6489361 | 73 | |
KC426930.1 | - | 12 - 13 | 1281 | 73 | ||
KC426931.1 | - | 12 - 13 | 1278 | 73 | ||
KC426932.1 | - | 11 - 12 | 1248 | 73 | ||
KC426933.1 | - | 11 - 12 | 1245 | 73 | ||
KC426934.1 | - | 11 - 12 | 1233 | 73 | ||
KC426935.1 | - | 11 - 12 | 1230 | 73 | ||
KC426936.1 | - | 10 - 11 | 1200 | 73 | ||
KC426937.1 | - | 10 - 11 | 1197 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
