Assay Details
Gene Symbol: Camsap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163749.1 | NP_001157221.1 | 14 - 15 | 3592 | 78 | |
NM_027171.3 | NP_081447.2 | 14 - 15 | 3589 | 78 | ||
GenBank mRNA | AK009960.1 | - | 2 - 3 | 176 | 78 | |
AK081728.1 | - | 14 - 15 | 3364 | 78 | ||
AK220401.1 | - | 9 - 10 | 3704 | 78 | ||
BC037466.1 | - | 4 - 5 | 1427 | 78 | ||
BC043033.1 | - | 6 - 7 | 2207 | 78 | ||
BC043081.1 | - | 14 - 15 | 3248 | 78 | ||
BC048787.1 | - | 14 - 15 | 3237 | 6489361 | 78 | |
KC426930.1 | - | 16 - 17 | 3311 | 78 | ||
KC426931.1 | - | 16 - 17 | 3308 | 78 | ||
KC426932.1 | - | 15 - 16 | 3278 | 78 | ||
KC426933.1 | - | 15 - 16 | 3275 | 78 | ||
KC426934.1 | - | 15 - 16 | 3263 | 78 | ||
KC426935.1 | - | 15 - 16 | 3260 | 78 | ||
KC426936.1 | - | 14 - 15 | 3230 | 78 | ||
KC426937.1 | - | 14 - 15 | 3227 | 78 | ||
KC426938.1 | - | 16 - 17 | 2063 | 78 | ||
KC426939.1 | - | 16 - 17 | 2060 | 78 | ||
KC426940.1 | - | 15 - 16 | 2030 | 78 | ||
KC426941.1 | - | 15 - 16 | 2027 | 78 | ||
KC426942.1 | - | 15 - 16 | 2015 | 78 | ||
KC426943.1 | - | 15 - 16 | 2012 | 78 | ||
KC426944.1 | - | 14 - 15 | 1982 | 78 | ||
KC426945.1 | - | 14 - 15 | 1979 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
