Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025768.2 | NP_080044.2 | 4 - 5 | 574 | 117 | |
GenBank mRNA | AK007905.1 | - | 4 - 5 | 579 | 117 | |
AK017197.1 | - | 4 - 5 | 538 | 117 | ||
AK044437.1 | - | 538 | 117 | |||
AK082863.1 | - | 4 - 5 | 568 | 117 | ||
AK149322.1 | - | 571 | 117 | |||
BC029096.1 | - | 4 - 5 | 580 | 3500563 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
